[(2R,3R,4S,5R,6R)-4,5-diacetoxy-6-bromo-2-[[tert-butyl(diphenyl)silyl]oxymethyl]tetrahydropyran-3-yl] acetate 1 * 1 g
Catalog # PHBLPBDL826-1G
Supplier: Pharmablock (USA)
[(2R,3R,4S,5R,6R)-4,5-diacetoxy-6-bromo-2-[[tert-butyl(diphenyl)silyl]oxymethyl]tetrahydropyran-3-yl] acetate 1 * 1 g
Catalog # PHBLPBDL826-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexane, EMPLURA®, Supelco®
Supplier: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, SupraSolv® for headspace gas chromatography, Supelco®
Supplier: Merck
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Butyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals