tert-butyl 7-(2-hydroxyethyl)-3,4-dihydro-2H-1,8-naphthyridine-1-carboxylate 1 * 1 g
Catalog # PHBLPBCH468-1G
Supplier: Pharmablock (USA)
tert-butyl 7-(2-hydroxyethyl)-3,4-dihydro-2H-1,8-naphthyridine-1-carboxylate 1 * 1 g
Catalog # PHBLPBCH468-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane 99+% for HPLC
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific