chloropalladium(1+) dicyclohexyl-[2-(2,4,6-triisopropylphenyl)phenyl]phosphane 2-phenylethanamine 1 * 1 g
Catalog # PHBLPBC00050-1G
Supplier: Pharmablock (USA)
chloropalladium(1+) dicyclohexyl-[2-(2,4,6-triisopropylphenyl)phenyl]phosphane 2-phenylethanamine 1 * 1 g
Catalog # PHBLPBC00050-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
Methanol dry (max. 0.005% water) , ACS, ISO 1 * 1 L
Supplier: PanReac AppliChem
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
Chloroform ≥99.0% stabilised BP, USP, NF for UV spectrophotometry, for IR spectrophotometry, for HPLC
Supplier: PanReac AppliChem
Acetone, meets analytical specification of Ph.Eur.
Supplier: Thermo Fisher Scientific
Chloroform stabilized with ethanol (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Methanol for UHPLC Supergradient, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Chloroform 99.8% stabilised for spectroscopy
Supplier: Thermo Fisher Scientific
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals