4-tert-butyl-2-(4-tert-butyl-2-pyridyl)pyridine 2-(2,4-difluorobenzene-6-id-1-yl)-5-(trifluoromethyl)pyridine iridium(3+) hexafluoro phosphate 1 * 1 g
Catalog # PHBLPBC00047-1G
Supplier: Pharmablock (USA)
4-tert-butyl-2-(4-tert-butyl-2-pyridyl)pyridine 2-(2,4-difluorobenzene-6-id-1-yl)-5-(trifluoromethyl)pyridine iridium(3+) hexafluoro phosphate 1 * 1 g
Catalog # PHBLPBC00047-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N-Bromosuccinimide 99%
Supplier: Thermo Fisher Scientific
1,4-Dioxane 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
Mineral oil light USP, NF, BP, Ph. Eur., pharma grade
Supplier: PanReac AppliChem
Methanol ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
n-Hexane ≥99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific