[2'-(amino-κN)[1,1'-biphenyl]-2-yl-κC][[2',6'-bis(1-methylethoxy)[1,1'-biphenyl]-2-yl]dicyclohexylphosphine-κP]chloro-Palladium 1 * 1 g
Catalog # PHBLPBC00032-1G
Supplier: Pharmablock (USA)
[2'-(amino-κN)[1,1'-biphenyl]-2-yl-κC][[2',6'-bis(1-methylethoxy)[1,1'-biphenyl]-2-yl]dicyclohexylphosphine-κP]chloro-Palladium 1 * 1 g
Catalog # PHBLPBC00032-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Toluene, anhydrous (max. 0.002% H₂O) ≥99.8%
Supplier: VWR Chemicals
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals