2-[4-[(2-bromophenoxy)methyl]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 1 * 1 g
Catalog # PHBLPBBD240-1G
Supplier: Pharmablock (USA)
2-[4-[(2-bromophenoxy)methyl]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 1 * 1 g
Catalog # PHBLPBBD240-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
2-Methylbutane for HPLC
Supplier: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, extra pure
Supplier: Thermo Fisher Scientific
Butyl acetate ≥99.5%, Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Trifluoroacetic acid 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific