2-[3-[(2-bromophenoxy)methyl]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 1 * 1 g
PHBLPBBD120-1G
2-[3-[(2-bromophenoxy)methyl]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 1 * 1 g
PHBLPBBD120-1G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals