5-[(3S)-5-fluoro-7-hydroxy-3-(isopentylamino)tetralin-6-yl]-1,1-dioxo-1,2,5-thiadiazolidin-3-one 1 * 100 mg
Catalog # PHBLPBB2019-100MG
Supplier: Pharmablock (USA)
5-[(3S)-5-fluoro-7-hydroxy-3-(isopentylamino)tetralin-6-yl]-1,1-dioxo-1,2,5-thiadiazolidin-3-one 1 * 100 mg
Catalog # PHBLPBB2019-100MG
Supplier: Pharmablock (USA)
Frequently Bought Together
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥98%, TECHNICAL
Supplier: VWR Chemicals
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
1,1,2,2-Tetrachloroethane 98.5%
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99%
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine solution 86-89% (by titrimetric analysis), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butanol 99+% for spectroscopy
Supplier: Thermo Fisher Scientific