O1-tert-butyl O2-methyl cis-4-(hydroxymethyl)pyrrolidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPB94533-1G
Supplier: Pharmablock (USA)
O1-tert-butyl O2-methyl cis-4-(hydroxymethyl)pyrrolidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPB94533-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Ethanolamine, EMSURE® for analysis, Supelco®
Supplier: Merck
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
2-Propanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methylcyclohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific