(1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxylic aci d 1 * 1 g
Catalog # PHBLPB93546-1G
Supplier: Pharmablock (USA)
(1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxylic aci d 1 * 1 g
Catalog # PHBLPB93546-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific
Sinapic acid, Supelco®
Supplier: Merck
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific