1-[(2R,3S,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione 1 * 1 g
Catalog # PHBLPB93409-1G
Supplier: Pharmablock (USA)
1-[(2R,3S,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione 1 * 1 g
Catalog # PHBLPB93409-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Acetonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Heptane, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
ISOPROPANOL 70% 1 * 1 L
Supplier: Biosepar
Dichloromethane 99.9% stabilised for residue analysis, for trace analysis of polyaromatic hydrocarbons
Supplier: Thermo Fisher Scientific
1-Pentanol 99%, pure
Supplier: Thermo Fisher Scientific