N-[3-(aminomethyl)pyrazin-2-yl]-N-methylmethanesulfonamide 1 * 1 g
Catalog # PHBLPB93200-1G
Supplier: Pharmablock (USA)
N-[3-(aminomethyl)pyrazin-2-yl]-N-methylmethanesulfonamide 1 * 1 g
Catalog # PHBLPB93200-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Pyridine, EMPLURA®, Supelco®
Supplier: Merck
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
n-Heptane
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Tetrahydrofuran stabilized with 300 ppm of BHT for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Methylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diiodomethane 99+% stabilised
Supplier: Thermo Fisher Scientific