tert-butyl 4-bromo-2-{1H-pyrrolo[2,3-b]pyridin-5-yloxy}benzoate 1 * 5 g
Catalog # PHBLPB93002-5G
Supplier: Pharmablock (USA)
tert-butyl 4-bromo-2-{1H-pyrrolo[2,3-b]pyridin-5-yloxy}benzoate 1 * 5 g
Catalog # PHBLPB93002-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
1,2,3,4-TETRAHYDRONAPHTHALENE FOR SYN... 1 * 200 kg
Supplier: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane, anhydrous 99+%
Supplier: Thermo Fisher Scientific