1-tert-butyl 2-methyl (2R,4R)-4-(hydroxymethyl)pyrrolidine-1,2-dicarboxylate 1 * 1 g
PHBLPB07731-1G
1-tert-butyl 2-methyl (2R,4R)-4-(hydroxymethyl)pyrrolidine-1,2-dicarboxylate 1 * 1 g
PHBLPB07731-1G
Frequently Bought Together
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
: PanReac AppliChem
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
: PanReac AppliChem
2-Propanol ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
1-Pentanol 99%, pure
: Thermo Fisher Scientific
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
: PanReac AppliChem
N,N-Dimethylacetamide 99%, pure
: Thermo Fisher Scientific
Methyl ethyl ketone ACS
: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
: Thermo Fisher Scientific
1-Pentanol 99+% ACS
: Thermo Fisher Scientific