tert-butyl 3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate 1 * 5 g
Catalog # PHBLPB05848-5G
Supplier: Pharmablock (USA)
tert-butyl 3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate 1 * 5 g
Catalog # PHBLPB05848-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals