tert-butyl cis-3-methoxy-4-(methylamino)piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPB05321-1G
Supplier: Pharmablock (USA)
tert-butyl cis-3-methoxy-4-(methylamino)piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPB05321-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals