tert-butyl rel-(3aR,5s,6aS)-5-amino-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-2-carboxylate hydrochloride 1 * 1 g
PHBLPB05177-01-1G
tert-butyl rel-(3aR,5s,6aS)-5-amino-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-2-carboxylate hydrochloride 1 * 1 g
PHBLPB05177-01-1G
Frequently Bought Together
2-Nitrophenylhydrazine, moistened 30% with water 97%
: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
: Honeywell Chemicals
Acetonitrile, extra dry 99.9%, AcroSeal®
: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
: Thermo Fisher Scientific
Isoamyl alcohol mixture of isomers ≥99%
: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals