tert-butyl cis-3-(aminomethyl)-4-hydroxypyrrolidine-1-carboxylate 1 * 1 g
Catalog # PHBLPB03139-1G
Supplier: Pharmablock (USA)
tert-butyl cis-3-(aminomethyl)-4-hydroxypyrrolidine-1-carboxylate 1 * 1 g
Catalog # PHBLPB03139-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals