tert-butyl 3-(methylamino)azetidine-1-carboxylate 1 * 5 g
Catalog # PHBLPB00239-5G
Leverancier: Pharmablock (USA)
tert-butyl 3-(methylamino)azetidine-1-carboxylate 1 * 5 g
Catalog # PHBLPB00239-5G
Leverancier: Pharmablock (USA)
Frequently Bought Together
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Leverancier: Merck
Ethylenediamine 99%
Leverancier: Thermo Fisher Scientific
2-Nitrophenylhydrazine, moistened 30% with water 97%
Leverancier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Leverancier: Honeywell Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Leverancier: Honeywell Chemicals
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Leverancier: Honeywell Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Leverancier: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Leverancier: PanReac AppliChem
Isobutyronitrile 99%
Leverancier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99%, pure
Leverancier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Leverancier: Roth Carl
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Leverancier: Honeywell Chemicals
Ethanol absolute, extra dry 99.5%, AcroSeal®
Leverancier: Thermo Fisher Scientific