3-(trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride 1 * 100 g
Catalog # PHBL00152-01-100G
Supplier: Pharmablock (USA)
3-(trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride 1 * 100 g
Catalog # PHBL00152-01-100G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
n-Heptane ≥99.3%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific