VIAL 40ML CLEAR SCREW 24MM 24-400 THREAD 28 x 95 MM 100/PACK 1 * 100 items
Catalog # HYPE6ASV40EPA-1
Supplier: Thermo Fisher Scientific
VIAL 40ML CLEAR SCREW 24MM 24-400 THREAD 28 x 95 MM 100/PACK 1 * 100 items
Catalog # HYPE6ASV40EPA-1
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Tubing connectors, b.safe
Supplier: Bohlender
Silikon/PTFE-Septa 1 * 100 ST
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
TSKgel® gel filtration columns, SW-type
Supplier: TOSOH Bioscience
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
Lids for flat bottom chambers
Supplier: CAMAG
Accessories for 7500/7700
Supplier: Agilent
Solid phase extraction cartridges, Bond Elut™ SI
Supplier: VARIAN
VWR®, Screw Caps for Screw Neck Vials ND18
Supplier: VWR Collection
Cross-disc inlet seals for Agilent GC's
Supplier: Restek
Microliter™ 700 series syringes
Supplier: HAMILTON BONADUZ