4-METHYLMORPHOLINE 4-OXIDE (50% SOLUTION IN WATER) FOR SYNTHESIS 1 * 1 Each
Catalog # 8.14779.9999
Supplier: Merck
4-METHYLMORPHOLINE 4-OXIDE (50% SOLUTION IN WATER) FOR SYNTHESIS 1 * 1 Each
Catalog # 8.14779.9999
Supplier: Merck
Frequently Bought Together
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
1-Butylboronic acid, Supelco®
Supplier: Merck