AD-MED VITRIGEL 2 (24 WELL)/PLATE 1 * 1 items
Catalogus # CHRA08364-96
Supplier: CHROMAGAR
Frequently Bought Together
ALLERGEN RAPID TEST EASY KIT: BUCKWHEAT 1 * 10 items
Supplier: HYSERVE
Corning® Trypsin
Supplier: Corning
Membrane filters with dehydrated media pads for microbiological control
Supplier: Sartorius
LID FOR STANDARD JAR 1 * 1 ST
Supplier: schuett-biotec GmbH
Dehydrated culture media
Supplier: LIOFILCHEM
SELECTIVE SUPPLEMENT MUP VIALS 1 * 10 items
Supplier: VWR Chemicals
Culture media, ready to use, in plates
Supplier: DIFCO BD
Antimicrobial susceptibility disks
Supplier: THERMO FISHER DIAGNOSTICS
PLATE COUNT AGAR 1 * 100 g
Supplier: LIOFILCHEM
CYCLOHEXIMIDE 0.1% SOLUTION 1 * 100 mL
Supplier: THERMO FISHER DIAGNOSTICS
Dehydrated Media
Supplier: THERMO FISHER DIAGNOSTICS
VWR®, Pre-weighed dehydrated culture media in sachets
Supplier: VWR Chemicals
SLANETZ BARTLEY AGAR 60MM PLATE 1 * 20 items
Supplier: LIOFILCHEM