COMPONENT 2 MIX 1 * 10 mL
Catalog # CMPLRD09490610
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Visual colour kits, sulphite, MQuant®, Supelco®
Supplier: Merck
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
VISOCOLOR® Colorimetric Tests, Chlorine
Supplier: MACHEREY-NAGEL
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
Test strips, peracetic acid, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, Chloride, LCK cuvette tests
Supplier: Hach