Order Entry
Netherlands
ContactUsLinkComponent
SHODEX GPC K-5004, PREPARATIVE SIZE EXCLUSION COLUMN, 50x300mm, 15µm, 1,500Å, CHLOROFORM 1 * 1 items
Catalog # SHOWF6109040
Supplier:  Resonac
SHODEX GPC K-5004, PREPARATIVE SIZE EXCLUSION COLUMN, 50x300mm, 15µm, 1,500Å, CHLOROFORM 1 * 1 items
Catalog # SHOWF6109040
Supplier:  Resonac
Frequently Bought Together