XSelect Peptide HSS T3 Column, 100Å, 5 µm, 2.1 mm X 150 mm, 1K - 15K 1 * 3 items
Catalog # WATE186008787
Supplier: WATERS
XSelect Peptide HSS T3 Column, 100Å, 5 µm, 2.1 mm X 150 mm, 1K - 15K 1 * 3 items
Catalog # WATE186008787
Supplier: WATERS
Frequently Bought Together
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
Vydac 218TP C18, HPLC columns
Supplier: Avantor
HPLC columns, ZORBAX® Eclipse XDB
Supplier: Agilent
Screw caps for screw neck vials, N 13
Supplier: MACHEREY-NAGEL
Capillary columns, Rtx®-1301
Supplier: Restek
Avantor® ACE® UltraCore BIO phenyl2, Analytical HPLC/UHPLC Columns
Supplier: Avantor
CrossLab supplies for GC systems: for PerkinElmer
Supplier: Agilent
Size exclusion chromatography columns, InfinityLab AdvanceBio
Supplier: Agilent
Capsule sinkers, SMI-LabHut
Supplier: QUALITY LAB ACCESSORIES
HPLC columns, PLRP-S
Supplier: VARIAN
HPLC columns, Hypersil™ BDS C18
Supplier: Thermo Fisher Scientific
VWR®, HPTLC Glass/Aluminium Plates, Silica, Modified Layers
Supplier: VWR Collection