BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 µm, 3 mm X 100 mm 1 * 1 items
Catalog # WATE186008949
Supplier: WATERS
BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 µm, 3 mm X 100 mm 1 * 1 items
Catalog # WATE186008949
Supplier: WATERS
Frequently Bought Together
Safety canisters
Supplier: S.C.A.T.
Gold standard syringes for autosampler
Supplier: Agilent
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Air valves, b.safe
Supplier: Bohlender
VWR®, Storage Boxes for Sample Vials/Bottles
Supplier: VWR Collection
Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
Split/splitless inlet seals
Supplier: Agilent
Screw caps for certified screw top vials
Supplier: Agilent
Safety canisters, electrically conducting
Supplier: S.C.A.T.
VWR® Standard Micro Syringes
Supplier: VWR Collection
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
Screw caps for screw neck vials, N 13
Supplier: MACHEREY-NAGEL
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
LC vials and closures
Supplier: Agilent
Headspace crimp caps with 20 mm septa
Supplier: Agilent