ACQUITY Premier Peptide HSS T3 Column, 100Å, 1.8 μm, 2.1 x 50 mm 1 * 1 items
Catalog # WATE186009490
Supplier: WATERS
ACQUITY Premier Peptide HSS T3 Column, 100Å, 1.8 μm, 2.1 x 50 mm 1 * 1 items
Catalog # WATE186009490
Supplier: WATERS
Frequently Bought Together
Crimp top vials and closures, 11 mm
Supplier: Agilent
Glass crimp top headspace vials, 20 ml, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Glass screw top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Vial Racks
Supplier: VWR Collection
Safety canisters
Supplier: S.C.A.T.
J.T.Baker® BAKERBOND spe™ Aromatic Sulphonic Acid, SPE Columns
Supplier: Avantor
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
TSKgel® gel filtration columns, SW-type
Supplier: TOSOH Bioscience
Inlet liners
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Tubing connectors, b.safe
Supplier: Bohlender
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR® Standard Micro Syringes
Supplier: VWR Collection
Canisters, GL 45, b.safe
Supplier: Bohlender
Stopcocks, three-way, Supelco®
Supplier: Merck