XSelect Premier Peptide CSH C18 Column, 130Å, 2.5 µm, 2.1 x 50 mm 1 * 1 items
Catalog # WATE186009904
Supplier: WATERS
XSelect Premier Peptide CSH C18 Column, 130Å, 2.5 µm, 2.1 x 50 mm 1 * 1 items
Catalog # WATE186009904
Supplier: WATERS
Frequently Bought Together
XCORE UHPLC Columns, C18
Supplier: ISERA
Inserts for crimp top and screw thread vials
Supplier: Restek
LiChrospher® RP-18E, HPLC Columns
Supplier: Avantor
SHUTOFF VALVE 1 * 1 items
Supplier: Agilent
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Capillary dispensers
Supplier: CAMAG
Silikon/PTFE-Septa, 1mm Dicke 1 * 100 ST
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
ISAspher Reversed Phase HPLC Columns, Amino
Supplier: ISERA
Torus DEA Column, 130Å, 5 µm, 4.6 mm X 50 mm 1 * 1 items
Supplier: WATERS
Ion chromatography columns, anion analysis
Supplier: Resonac
HSPgel Column, 3 µm, 6 mm X 150 mm, 500 - 10K 1 * 1 items
Supplier: WATERS
Gloves
Supplier: Agilent