PoraPak Rxn RP 6 cc Vac Cartridge, 400 mg Sorbent per Cartridge, 80 µm 1 * 30 items
Catalog # WATE186004545
Supplier: WATERS
PoraPak Rxn RP 6 cc Vac Cartridge, 400 mg Sorbent per Cartridge, 80 µm 1 * 30 items
Catalog # WATE186004545
Supplier: WATERS
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR® Standard Micro Syringes
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Snap caps for snap cap vials, N 18 and N 22
Supplier: MACHEREY-NAGEL
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection