PoraPak Rxn CX 6 cc Flangeless Vac Cartridge, 400 mg Sorbent per Cartridge, 80 µm 1 * 30 items
Catalog # WATE186004542
Supplier: WATERS
PoraPak Rxn CX 6 cc Flangeless Vac Cartridge, 400 mg Sorbent per Cartridge, 80 µm 1 * 30 items
Catalog # WATE186004542
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® Generix® C18 & C18(2), Analytical HPLC Columns
Supplier: Avantor
ISAspher HPLC Columns, Silica, HILIC
Supplier: ISERA
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
FlashPure EcoFlex Silica Cartridges
Supplier: BUCHI
GC columns, Ultra Inert, J&W DB-WAX
Supplier: Agilent
Screw caps, 9 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
Accessories for flame ionization detector (FID), auto-ignite
Supplier: PerkinElmer
Hose connectors, b.safe
Supplier: Bohlender
HPLC column tubing
Supplier: Restek
HPLC columns, InfinityLab Poroshell 120 HPH-C18
Supplier: Agilent
L-MARK® Replacement Plunger for CTC Analytics PAL-xt
Supplier: Trajan Scientific and Medical
Diamond RSH/RTC syringes
Supplier: Trajan Scientific and Medical
500UL SAMPLE LOOP 1 * 1 items
Supplier: PerkinElmer
LC vials and closures
Supplier: Agilent
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ