Sep-Pak Florisil 1 cc Vac Cartridge, 100 mg Sorbent per Cartridge, 50 - 200 µm, 1 * 100 items
Catalog # WATEWAT023600
Supplier: WATERS
Sep-Pak Florisil 1 cc Vac Cartridge, 100 mg Sorbent per Cartridge, 50 - 200 µm, 1 * 100 items
Catalog # WATEWAT023600
Supplier: WATERS
Frequently Bought Together
Microliter™ 700 series syringes
Supplier: HAMILTON BONADUZ
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
TSKgel® gel filtration columns, SW-type
Supplier: TOSOH Bioscience
Tubing connectors, b.safe
Supplier: Bohlender
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Screw caps for screw neck vials, N 24
Supplier: MACHEREY-NAGEL
VWR®, Screw Caps for Screw Neck Vials ND10
Supplier: VWR Collection
Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
Accessories for TLC sprayer
Supplier: Merck
Spouts for jerricans
Supplier: S.C.A.T.
HPLC columns, InfinityLab Poroshell 120 HPH-C18
Supplier: Agilent
Avantor® Alltima, HPLC Columns, C18-LL
Supplier: Avantor
Capillary columns, Rtx®-5MS (fused silica)
Supplier: Restek
AccQ-Tag Ultra - Derivitization Kit (250 analyses) 1 * 1 KIT
Supplier: WATERS
VWR® Hichrom C8 HPLC Columns
Supplier: Avantor