CORTECS C8 Method Validation Kit, 90Å, 1.6 µm, 3 mm X 150 mm 1 * 3 items
Catalogus # WATE186008417
Supplier: WATERS
CORTECS C8 Method Validation Kit, 90Å, 1.6 µm, 3 mm X 150 mm 1 * 3 items
Catalogus # WATE186008417
Supplier: WATERS
Frequently Bought Together
RCM 8 X 10 EXTENSION KIT 1 * 1 KIT
Supplier: WATERS
CHROMABOND® SiOH SPE Columns
Supplier: MACHEREY-NAGEL
FILAMENT BLOCK 1 * 1 items
Supplier: Agilent
SGE SolGel WAX™ GC Capillary Columns
Supplier: Trajan Scientific and Medical
DISCOVERY DSI-70 PK/54 3ML TUBE 500MG 1 * 54 items
Supplier: Merck
Hollow cathode lamps, Lumina™
Supplier: PerkinElmer
Avantor® ACE® AQ, HPLC Columns, 5 µm
Supplier: Avantor
HPLC columns, NUCLEOSIL® SB
Supplier: MACHEREY-NAGEL