ACQUITY UPLC HSS C18 Method Validation Kit, 100Å, 1.8 µm, 2.1 mm X 100 mm 1 * 3 items
Catalog # WATE186004058
Supplier: WATERS
ACQUITY UPLC HSS C18 Method Validation Kit, 100Å, 1.8 µm, 2.1 mm X 100 mm 1 * 3 items
Catalog # WATE186004058
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C18-HL, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
TLC plates, silica gel 60, unmodified layers, SIL G
Supplier: MACHEREY-NAGEL
Avantor® ACE® UltraCore BIO, Analytical HPLC/UHPLC Columns
Supplier: Avantor
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
HPLC columns, Poroshell 120 EC-C18
Supplier: Agilent
Avantor® Partisil SAX, HPLC Columns
Supplier: Avantor
PK20 PRECLEANED ASSEMBLED 1/4IN BRASS 1 * 1 items
Supplier: Merck
HPLC columns, PLRP-S
Supplier: VARIAN
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
HPLC columns, SUPELCOSIL™, LC-8-DB, Supelco®
Supplier: Merck
Crimp Caps for Crimp Neck Vials, N 11
Supplier: MACHEREY-NAGEL
LC vials and closures
Supplier: Agilent
ISAspher HPLC Columns, C18 BDS
Supplier: ISERA