CORTECS Phenyl Method Validation Kit, 90Å, 1.6 µm, 2.1 mm X 100 mm 1 * 3 items
Catalog # WATE186008386
Supplier: WATERS
CORTECS Phenyl Method Validation Kit, 90Å, 1.6 µm, 2.1 mm X 100 mm 1 * 3 items
Catalog # WATE186008386
Supplier: WATERS
Frequently Bought Together
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
J.T.Baker® BAKERBOND spe™ SiOH, SPE Columns
Supplier: Avantor
Safety canisters
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Manual crimping tools, advanced ergonomic version
Supplier: MACHEREY-NAGEL
Spouts, b.safe
Supplier: Bohlender
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Supplier: PerkinElmer