XBridge BEH C8 Method Validation Kit, 130Å, 5 µm, 3 mm X 100 mm 1 * 3 items
Catalog # WATE186003783
Supplier: WATERS
XBridge BEH C8 Method Validation Kit, 130Å, 5 µm, 3 mm X 100 mm 1 * 3 items
Catalog # WATE186003783
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
SymLine Automatic Switching for Container
Supplier: S.C.A.T.
[EN]SEPTA THERMOLITE PLUS SHIMADZU PLUG 1 * 100 ST
Supplier: Restek
DHA Olefins standard
Supplier: Restek
SEC standards, AdvanceBio
Supplier: Agilent
HPLC columns, Hypersil™ BDS C8
Supplier: Thermo Fisher Scientific
TLC plates, silica gel 60, unmodified layers, SIL G
Supplier: MACHEREY-NAGEL
Vydac 214TP C4, HPLC columns
Supplier: Avantor