XSelect HSS T3 Method Validation Kit, 100Å, 2.5 µm, 2.1 mm X 150 mm 1 * 3 items
Catalog # WATE186006797
Supplier: WATERS
XSelect HSS T3 Method Validation Kit, 100Å, 2.5 µm, 2.1 mm X 150 mm 1 * 3 items
Catalog # WATE186006797
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C18-HL, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
VWR®, Septa GL 45
Supplier: VWR Collection
VWR® Hitachi LaChromUltra II, LaChrom HPLC Columns
Supplier: VWR Collection
Avantor® ACE® C18-AR, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Snap Cap Vials
Supplier: VWR Collection
FlashPure Select Silica Cartridges
Supplier: BUCHI
Microliter™ and Gastight® syringes for Waters® U6K injection valve
Supplier: HAMILTON BONADUZ
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Avantor® Alltima, HPLC Columns, C8
Supplier: Avantor
CHIRALPAK® IH 250 x 20 mm 5 µm SFC Semi-Prep Column 1 * 1 items
Supplier: CHIRAL TECHNOLOGIES
GC capillary column connectors, Swagelok®
Supplier: PerkinElmer
ISAspher HPLC Columns, C18 BDS
Supplier: ISERA