CORTECS C8 Method Validation Kit, 90Å, 2.7 µm, 4.6 mm X 150 mm 1 * 3 items
Catalog # WATE186008377
Supplier: WATERS
CORTECS C8 Method Validation Kit, 90Å, 2.7 µm, 4.6 mm X 150 mm 1 * 3 items
Catalog # WATE186008377
Supplier: WATERS
Frequently Bought Together
TSKgel® gel filtration columns, SW-type
Supplier: TOSOH Bioscience
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE®, Guard Cartridge Holders
Supplier: Avantor
Glass screw top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Geosmin and 2-methylisoborneol P&TM, 1mL, 100ug/mL 1 * 1 Ampoul
Supplier: Restek
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
Swabs, cotton
Supplier: Agilent
Snap top vials and closures, 11 mm
Supplier: Agilent
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Accessories for GC/MS
Supplier: Agilent
Microliter™ 700 series syringes
Supplier: HAMILTON BONADUZ
Valves for syringes with valves
Supplier: Trajan Scientific and Medical
FlashPure Select Silica Cartridges
Supplier: BUCHI
SPME fiber assemblies, Supelco®
Supplier: Merck
HPTLC Plates, Silica Gel 60, Modified Layers, RP-18 W/UV₂₅₄
Supplier: MACHEREY-NAGEL
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Supplier: PerkinElmer