XSelect Peptide CSH C18 Method Validation Kit, 130Å, 3.5 µm, 4.6 mm X 100 mm, 1K - 15K 1 * 3 items
Catalog # WATE186006959
Supplier: WATERS
XSelect Peptide CSH C18 Method Validation Kit, 130Å, 3.5 µm, 4.6 mm X 100 mm, 1K - 15K 1 * 3 items
Catalog # WATE186006959
Supplier: WATERS
Frequently Bought Together
Screw neck vials, N 18 and N 20
Supplier: MACHEREY-NAGEL
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Safety canisters
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
HPLC columns, LiChrospher®
Supplier: Merck
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
Certified screw top vials and closures
Supplier: Agilent
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Sample vials, E-C, WHEATON®
Supplier: DWK Life Sciences
Screw caps for screw neck vials, N 13
Supplier: MACHEREY-NAGEL
LC vials and closures
Supplier: Agilent