XBridge BEH C8 Method Validation Kit, 130Å, 3.5 µm, 4.6 mm X 100 mm 1 * 3 items
Catalog # WATE186003780
Supplier: WATERS
XBridge BEH C8 Method Validation Kit, 130Å, 3.5 µm, 4.6 mm X 100 mm 1 * 3 items
Catalog # WATE186003780
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
COLUMN 130Å 2.5 µM C18 XBRIDGE BEH XP 1 * 1 items
Supplier: WATERS
ISAspher HPLC Columns, C8
Supplier: ISERA
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Size exclusion chromatography columns, Bio SEC-3 and Bio SEC-5
Supplier: Agilent
LC vials and closures
Supplier: Agilent
Metab-AAC HPLC Columns, BF Series
Supplier: ISERA
Canisters, S60/61, with level indicator, b.safe
Supplier: Bohlender
Tubing, PTFE
Supplier: HAMILTON BONADUZ
Avantor® ACE® Excel® C18, HPLC/UHPLC Columns, Analytical, 1,7 µm
Supplier: Avantor
Sample Loops for Rheodyne® Valves
Supplier: Agilent
Bulk media for HPLC, POLYGOPREP
Supplier: MACHEREY-NAGEL
HPLC column tubing
Supplier: Restek
SunFire C8 Column, 100Å, 5 µm, 2.1 mm X 50 mm 1 * 1 items
Supplier: WATERS