ACQUITY UPLC M-Class Peptide BEH C18 Column, 300Å, 1.7 µm, 100 µm X 100 mm 1 * 1 items
WATE186007488
: WATERS
ACQUITY UPLC M-Class Peptide BEH C18 Column, 300Å, 1.7 µm, 100 µm X 100 mm 1 * 1 items
WATE186007488
: WATERS
Frequently Bought Together
VWR®, Screw Caps for Screw Neck Vials ND24
: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
: Avantor
VWR®, Short Thread Vials ND9
: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
: VWR Collection
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
: MERCK PRODUCTION CHEMICALS
VWR®, Inserts for Short Thread Vials ND9
: VWR Collection
VWR®, Screw Neck Vials ND13
: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
: VWR Collection
VWR®, Crimp Neck Vials ND11
: VWR Collection
Florisil® 60-100 mesh for chromatography
: VWR Chemicals
Headspace crimp caps with 20 mm septa
: Agilent
VWR®, Snap Cap Vials
: VWR Collection
Exhaust filters, V3.0
: S.C.A.T.