XSelect CSH C18 OBD Prep Column, 130Å, 5 µm, 50 mm X 250 mm 1 * 1 items
Catalog # WATE186005497
Supplier: WATERS
XSelect CSH C18 OBD Prep Column, 130Å, 5 µm, 50 mm X 250 mm 1 * 1 items
Catalog # WATE186005497
Supplier: WATERS
Frequently Bought Together
Microlitre™ syringes for manual HPLC injections on Rheodyne and Valco valves
Supplier: HAMILTON BONADUZ
Safety canisters
Supplier: S.C.A.T.
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
TSKgel® gel filtration columns, SW-type
Supplier: TOSOH Bioscience
Inlet liners
Supplier: Agilent
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Accessories for GC/MS
Supplier: Agilent
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
Inlet septa
Supplier: Agilent
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
N 20 stopper Bromobutyl, gray null
Supplier: MACHEREY-NAGEL
FlashPure EcoFlex Silica Cartridges
Supplier: BUCHI