CORTECS HILIC Column, 90Å, 2.7 µm, 4.6 mm X 150 mm 1 * 3 items
Catalog # WATE176003321
Supplier: WATERS
CORTECS HILIC Column, 90Å, 2.7 µm, 4.6 mm X 150 mm 1 * 3 items
Catalog # WATE176003321
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
Gastight® 1800 series syringes
Supplier: HAMILTON BONADUZ
Safety Waste Cap "LISA", V3.0
Supplier: S.C.A.T.
Gas purifiers/filters, Big Trap
Supplier: Trajan Scientific and Medical
Chromatography Tubing, Stainless Steel
Supplier: Avantor
SafetyWasteCaps
Supplier: S.C.A.T.
SEAT ASSEMBLY PEEK 0.12MM 1 * 1 items
Supplier: Agilent
Traps for turbomatrix thermal desorbers
Supplier: PerkinElmer
Size exclusion chromatography columns, GPC KF-800 series
Supplier: Resonac
GC columns, Ultra Inert, J&W DB-WAX
Supplier: Agilent