XBridge BEH C8 OBD Prep Column, 130Å, 5 µm, 19 mm X 150 mm 1 * 1 items
Catalog # WATE186002995
Supplier: WATERS
XBridge BEH C8 OBD Prep Column, 130Å, 5 µm, 19 mm X 150 mm 1 * 1 items
Catalog # WATE186002995
Supplier: WATERS
Frequently Bought Together
Certified screw top vials and closures
Supplier: Agilent
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Vial Racks
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Gold standard syringes for autosampler
Supplier: Agilent
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT
Microvolume syringes 10 µl
Supplier: Trajan Scientific and Medical