Order Entry
Netherlands
ContactUsLinkComponent
CORTECS C18+ Column, 90Å, 1.6 µm, 2.1 mm X 100 mm 1 * 3 items
  WATE176003169
 :  WATERS
CORTECS C18+ Column, 90Å, 1.6 µm, 2.1 mm X 100 mm 1 * 3 items
  WATE176003169
 :  WATERS
Frequently Bought Together