ACQUITY UPLC BEH C18 Column, 130Å, 1.7 µm, 3 mm X 150 mm 1 * 3 items
Catalog # WATE176001797
Supplier: WATERS
ACQUITY UPLC BEH C18 Column, 130Å, 1.7 µm, 3 mm X 150 mm 1 * 3 items
Catalog # WATE176001797
Supplier: WATERS
Frequently Bought Together
Headspace screw neck vials, N 18
Supplier: MACHEREY-NAGEL
Certified screw top vials and closures
Supplier: Agilent
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
SafetyWasteCaps
Supplier: S.C.A.T.
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Avantor® Ultrasphere®, HPLC Columns
Supplier: Avantor
SafetyCaps
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 1,7 µm
Supplier: Avantor
Avantor® ACE® C8, HPLC Columns, 3 µm
Supplier: Avantor
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Crimp caps for crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Capillary column ferrules for gas chromatography
Supplier: Agilent
Supplies for pumps
Supplier: Agilent
Seals and closures for 8 mm screw vials, Chromacol™
Supplier: CHROMACOL