Symmetry Shield RP18 Prep Column, 100Å, 5 µm, 19 mm X 100 mm 1 * 1 items
Catalog # WATE186001837
Supplier: WATERS
Symmetry Shield RP18 Prep Column, 100Å, 5 µm, 19 mm X 100 mm 1 * 1 items
Catalog # WATE186001837
Supplier: WATERS
Frequently Bought Together
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR® Air Valve
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Flat bottom chambers
Supplier: CAMAG