CORTECS Phenyl Column, 90Å, 2.7 µm, 2.1 mm X 50 mm 1 * 3 items
Catalog # WATE176003790
Supplier: WATERS
CORTECS Phenyl Column, 90Å, 2.7 µm, 2.1 mm X 50 mm 1 * 3 items
Catalog # WATE176003790
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Certified headspace crimp top vials
Supplier: Agilent
HPLC columns, LiChrospher®
Supplier: Merck
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR® Air Valve
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection