CORTECS C18 Column, 90Å, 2.7 µm, 2.1 mm X 50 mm 1 * 3 items
Catalog # WATE176003270
Supplier: WATERS
CORTECS C18 Column, 90Å, 2.7 µm, 2.1 mm X 50 mm 1 * 3 items
Catalog # WATE176003270
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® Excel® C18-AR, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
L-MARK® LC Autosampler Syringes
Supplier: Trajan Scientific and Medical
XSelect CSH C18 Column, 130Å, 3.5 µm, 3 mm X 30 mm 1 * 1 items
Supplier: WATERS
Consumables for Thermo Scientific autosamplers
Supplier: Trajan Scientific and Medical
Replacement cap frit filters for Trident guard cartridges
Supplier: Restek
Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
TUBES TOP-DNA 150MG 20 X 1 * 96 items
Supplier: VARIAN
Discovery® Classical C18 HPLC Columns
Supplier: Merck
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Mini-display table level sensor, b.safe
Supplier: Bohlender
500UL SAMPLE LOOP 1 * 1 items
Supplier: PerkinElmer
ISAspher HPLC Columns, C18-AQ-PolE
Supplier: ISERA