XSelect Peptide CSH C18 OBD Prep Column, 130Å, 5 µm, 19 mm X 50 mm, 1K - 15K 1 * 1 items
Catalog # WATE186007022
Supplier: WATERS
XSelect Peptide CSH C18 OBD Prep Column, 130Å, 5 µm, 19 mm X 50 mm, 1K - 15K 1 * 1 items
Catalog # WATE186007022
Supplier: WATERS
Frequently Bought Together
HPLC Columns, Reprospher 100 C18
Supplier: DR MAISCH
Solid phase extraction cartridges, Bond Elut™ C18
Supplier: VARIAN
Achiral analytical dcpak® pmpc 150x2.1mm 3 micron 1 * 1 items
Supplier: CHIRAL TECHNOLOGIES
Capillary columns, CP-Sil
Supplier: VARIAN
Waste Tubing Desktop and Rear Mounts, SymLine®
Supplier: S.C.A.T.
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
XTerra MS C8 Column, 125Å, 3.5 µm, 4.6 mm X 150 mm 1 * 1 items
Supplier: WATERS
GC COLUMN HI-1 0.10MM 0.10UM 20M 1 * 1 items
Supplier: Avantor
Avantor® ACE® UltraCore BIO phenyl2, Analytical HPLC/UHPLC Columns
Supplier: Avantor